Learn More
Z-Lys(z)-osu, TRC
Product Code. 30477150
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30477150 100 mg 100mg
30483391 250 mg 250mg
30474346 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30477150 Supplier Toronto Research Chemicals Supplier No. TRCL488848100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21160-83-8
Molecular Formula C26 H29 N3 O8
Molecular Weight (g/mol) 511.52
Synonym L-Lysine, N2,N6-bis[(phenylmethoxy)carbonyl]-, 2,5-dioxo-1-pyrrolidinyl ester, Carbamic acid, [(1S)-1-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]-1,5-pentanediyl]bis-, bis(phenylmethyl) ester (9CI), Carbamic acid, [1-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]-1,5-pentanediyl]bis-, bis(phenylmethyl) ester, (S)-, Succinimide, N-[(N2,N6-dicarboxy-L-lysyl)oxy]-, dibenzyl ester (8CI), NSC 250409, Nα,Nε-Dibenzyloxycarbonyl-L-lysine N-hydroxysuccinimide ester
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (2S)-2,6-bis(phenylmethoxycarbonylamino)hexanoate
SMILES O=C(NCCCC[C@H](NC(=O)OCc1ccccc1)C(=O)ON2C(=O)CCC2=O)OCc3ccccc3
Specifications
InChI Formula InChI=1S/C26H29N3O8/c30-22-14-15-23(31)29(22)37-24(32)21(28-26(34)36-18-20-11-5-2-6-12-20)13-7-8-16-27-25(33)35-17-19-9-3-1-4-10-19/h1-6,9-12,21H,7-8,13-18H2,(H,27,33)(H,28,34)/t21-/m0/s1
Quantity 100 mg
Formula Weight 511.1955
Shipping Condition Room Temperature
Chemical Name or Material 2,5-Dioxopyrrolidin-1-yl (2S)-2,6-Bis(benzyloxycarbonylamino)hexanoate