Learn More
MedChemExpress Z-Gly-Gly-Arg-AMC acetate
Specifications
Specifications
| CAS | 2070009-61-7 |
| Solubility | DMSO : 500 mg/mL (781.67 mM∣Need ultrasonic) ∣H2O : 20 mg/mL (31.27 mM∣Need ultrasonic) |
| Content And Storage | Sealed storage, away from moisture and light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light) |
| Quantity | 5 mg |
| Molecular Weight (g/mol) | 639.66 g/mol |
| Product Type | Dye Reagents |
| Purity or Quality Grade | 98.02% |
| Shipping Condition | Room temperature |