Learn More
MedChemExpress Z-Gly-Gly-Arg-AMC acetate
Product Code. 16419328
Encompass_Preferred
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 16419328
Brand: MedChemExpress HYP0019A10mg
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Z-Gly-Gly-Arg-AMC acetate is a thrombin-specific fluorogenic substrate for testing of thrombin generation in PRP and platelet-poor plasma (PPP).
Specifications
CAS 2070009-61-7
Solubility DMSO : 500 mg/mL (781.67 mM∣Need ultrasonic) ∣H2O : 20 mg/mL (31.27 mM∣Need ultrasonic)
Content And Storage Sealed storage, away from moisture and light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
Quantity 10 mg
Molecular Weight (g/mol) 639.66 g/mol
Product Type Dye Reagents
Purity or Quality Grade 98.02%
Shipping Condition Room temperature