missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Z 944

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Z 944 is a selective T-type Ca2+ channel blocker (IC50 values are 50 to 160 nM for hCav3.1, hCav3.2 and hCav3.3). Exhibits 50 to 600 fold selectivity for Cav3.x channels over related ion channels (Cav1.2, hERG and Nav1.5). Has no effect on action potential properties in isolated heart Purkinje fibers and no adverse effects on ECG parameters in vivo. Exhibits anti-seizure effects in animal models.
Specifications
Specifications
| Chemical Name or Material | 4-[[(3-Chloro-5-fluorobenzoyl)amino)methyl]-N-(1,1-dimethylethyl)-1-piperidineacetamide |
| CAS | 1199236-64-0 |
| Quantity | 10 mg |
| Molecular Formula | C19H27ClFN3O2 |
| Formula Weight | 383.89 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction