missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies (Z)-4-Hydroxytamoxifen

Product Code. p-7217498
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 17100322

Brand: STEMCELL Technologies 74056

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Estrogen signaling pathway modulator; Modulates selective estrogen receptor and estrogen-related receptor

  • (Z)-4-Hydroxytamoxifen is a cell-permeable selective estrogen receptor modulator (SERM) that binds to estrogen receptors (ER) and estrogen-related receptors (ERR) with both estrogenic and anti-estrogenic effects (Poulin et al.)
  • Suppresses tumor growth in breast cancer cells (Freiss et al.; Osborne et al.)

Specifications

CAS 68047-06-3
Synonym trans-4-Hydroxytamoxifen; 4-OH-TAM; ICI 79280
Quantity 25 mg
Molecular Formula C26H29NO2
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.