Learn More
Fisherbrand™ Plastic Radiation Hazard Trays
Product Code. 12357247 Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
46 x 26cm
57 x 54cm
113 x 54cm
Unit Size:
Each
Product Code. 12357247
Please to purchase this item. Need a web account? Register with us today!
Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.
  • Radiation hazard tray yellow 113x54cm
  • Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
  • All trays supplied with one free liner when purchased
  • Additional environmentally friendly APET liners are provided in packs of 25
Specifications
Dimensions (L x W) 113 x 54cm
Color Yellow
Material Plastic
Type Radiation Hazard Tray
For Use With Safe containment of spillages, lab bench protection
Includes Liner
No. per Pack 1 Each
Shape Rectangular