Learn More
Ibidi Y Tube Fitting
Product Code. 16833963
Encompass_Preferred
Click to view available options
Diameter (Metric) Inner:
0.8 mm
1.6 mm
Unit Size:
Pack of 25
This item is not returnable. View return policy
Product Code. 16833963
Brand: Ibidi 10827
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Y Tube Fitting is suitable for flexible tubes.
  • Suitable for flexible tubes
  • natural Polypropylene
  • sterilized
Specifications
Diameter (Metric) Inner 0.8 mm
Material Polypropylene
Packaging Quantity 25 Pk.
Sterility Sterile
Color Clear
Product Type Y Connector