Learn More
XRK3F2, MedChemExpress
Description
XRK3F2 is an inhibitor of p62 (Sequestosome-1)-ZZ/ domain.
Specifications
Specifications
| Chemical Name or Material | XRK3F2 |
| CAS | 2375193-43-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H24ClF2NO3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 150 mg/mL (344.12 mM; Need ultrasonic) ∣H2O : 0.91 mg/mL (2.09 mM; Need ultrasonic) |
| SMILES | FC(C=C1)=CC=C1COC2=C(OCC3=CC=C(F)C=C3)C=C(CNCCO)C=C2.Cl |
| Molecular Weight (g/mol) | 435.89 |
| Formula Weight | 435.89 |
| Show More |