missing translation for 'onlineSavingsMsg'
Learn More

XRCC6BP1 Mouse anti-Human, Clone: OTI5F3, TrueMAB™, Origene

Product Code. 17569593
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
Unit Size:
30µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17569593 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17569593 Supplier Origene Technologies Supplier No. TA808393S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

XRCC6BP1 Monoclonal antibody specifically detects XRCC6BP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen XRCC6BP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5F3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ATP23
Gene Accession No. Q9Y6H3
Gene Alias 1110068E08Rik; 2410012H02Rik; atp23; ATP23 metallopeptidase and ATP synthase assembly factor homolog; Ku70 binding protein 3; Ku70-binding protein 3; KUB3; LOW QUALITY PROTEIN: mitochondrial inner membrane protease ATP23 homolog; mitochondrial inner membrane protease ATP23 homolog; XRCC6 binding protein 1; XRCC6-binding protein 1; Xrcc6bp1; zgc:162885
Host Species Mouse
Immunogen Full length human recombinant protein of human XRCC6BP1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91419
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.