Learn More
XRCC1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-202PE
Product Code. 18502002
  • 615.00 € / 0.10mL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
XRCC1 Polyclonal antibody specifically detects XRCC1 in Human samples. It is validated for Western Blot, KnockDown