Learn More
XMD17-109, MedChemExpress
Product Code. 16422817
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422817 50 mg 50mg
16409287 5 mg 5mg
16406738 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16422817 Supplier Medchem Express Supplier No. HY15665
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress XMD17-109 is a novel, specific ERK-5 inhibitor, with an IC50 of 162 nM.
XMD17-109 is a novel, specific ERK-5 inhibitor, with an IC50 of 162 nM.
Specifications
Chemical Name or Material XMD17-109
CAS 1435488-37-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C36H46N8O3
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (156.54 mM)
SMILES O=C1C2=C(C=CC=C2)N(C3CCCC3)C4=NC(NC5=C(OCC)C=C(C(N6CCC(N7CCN(C)CC7)CC6)=O)C=C5)=NC=C4N1C
Molecular Weight (g/mol) 638.8
Formula Weight 638.8
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease