missing translation for 'onlineSavingsMsg'
Learn More

XMD16-5, MedChemExpress

Product Code. 16429846
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16429846

Brand: Medchem Express HY101243

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress XMD16-5 is a potent TNK2 inhibitor with IC50 values of 16 and 77 nM for the D163E and R806Q mutations, respectively.

XMD16-5 is a potent TNK2 inhibitor with IC50 values of 16 and 77 nM for the D163E and R806Q mutations, respectively.

Specifications

Chemical Name or Material XMD16-5
Physical Form Solid
CAS 1345098-78-3
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C23H24N6O2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 100 mg/mL (240.11 mM; Need ultrasonic)
SMILES O=C1NC2=CN=C(NC3=CC=C(N4CCC(O)CC4)C=C3)N=C2N(C)C5=CC=CC=C15
Molecular Weight (g/mol) 416.48
Formula Weight 416.48
Percent Purity 98.73%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.