missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XLF Antibody (OTI1B5), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74880AF405
This item is not returnable.
View return policy
Description
XLF Monoclonal specifically detects XLF in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| XLF | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NHEJ1 | |
| Full length recombinant protein of human XLF (NP_079058) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1B5 | |
| Western Blot, Immunohistochemistry | |
| Cernunnos, non-homologous end-joining factor 1, nonhomologous end-joining factor 1, Protein cernunnos, XLFFLJ12610, XRCC4-like factor | |
| Mouse | |
| Affinity Purified | |
| Cancer, Chromatin Research, DNA Repair | |
| 79840.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction