missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XLF Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-53799
This item is not returnable.
View return policy
Description
XLF Polyclonal specifically detects XLF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| XLF | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Cernunnos, non-homologous end-joining factor 1, nonhomologous end-joining factor 1, Protein cernunnos, XLFFLJ12610, XRCC4-like factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Expected from sequence similarity: Dog. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, Immunohistochemistry 2.5 - 3.8 ug/ml, Immunohistochemistry-Paraffin 2.5 - 3.8 ug/ml, Peptide ELISA Detection limit 1:64000 | |
| NP_079058.1 | |
| NHEJ1 | |
| Peptide with sequence C-QRPQLSKVKRKKPR corresponding to C-Terminus according to NP_079058.1. | |
| 0.1 mg | |
| Cancer, Chromatin Research, DNA Repair | |
| 79840 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction