missing translation for 'onlineSavingsMsg'
Learn More

XCT790, MedChemExpress

Product Code. 16490317
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16490317

Brand: Medchem Express HY10426

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress XCT-790 is a potent and selective inverse agonist for ERRα with an IC50 value of 0.37 μM. XCT-790 induces cell death in chemotherapeutic resistant cancer cells. XCT-790 (Compound 12) is inactive against ERRγ and the estrogen receptors ERα and ERβ.

XCT-790 is a potent and selective inverse agonist for ERRα with an IC50 value of 0.37 μM. XCT-790 induces cell death in chemotherapeutic resistant cancer cells. XCT-790 (Compound 12) is inactive against ERRγ and the estrogen receptors ERα and ERβ.

Specifications

Chemical Name or Material XCT790
CAS 725247-18-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H13F9N4O3S
Quantity 50 mg
Solubility Information DMSO : 16.67 mg/mL (27.95 mM; Need ultrasonic)
SMILES O=C(NC1=NN=C(C(F)(F)F)S1)/C(C#N)=C/C2=CC=C(OCC3=C(C(F)(F)F)C=C(C(F)(F)F)C=C3)C(OC)=C2
Molecular Weight (g/mol) 596.42
Formula Weight 596.42
Percent Purity 96.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.