Learn More
Tocris Bioscience™ XAP 044
GREENER_CHOICE
Potent and selective mGlu7 antagonist
Brand:  Tocris Bioscience™ 5248/50
Encompass
Product Code. 15766616
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
XAP 044 is a potent and selective mGlu7 antagonist (IC50 = 88 nM); inhibits lateral amygdala long term potentiation (LTP) in brain slices from wild type mice. Displays no effects on the LTP of mGlu7 deficient mice. Exhibits novel activity by binding to the mGlu7 Venus flytrap domain (VFTD). Exhibits anti-anxiety effects in rodent models. Brain penetrant.
Specifications
Specifications
>98%