Learn More
Xanthinol Nicotinate, TRC
Product Code. 30451395
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451395 10 mg 10mg
30454213 1 mg 1mg
30453233 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30451395 Supplier Toronto Research Chemicals Supplier No. TRCX49996010MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 437-74-1
Molecular Formula C13 H21 N5 O4 . C6 H5 N O2
Molecular Weight (g/mol) 434.45
Synonym 3-Pyridinecarboxylic acid, compd. with 3,7-dihydro-7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-1,3-dimethyl-1H-purine-2,6-dione (1:1) (9CI, ACI), 1H-Purine-2,6-dione, 3,7-dihydro-7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-1,3-dimethyl-, mono-3-pyridinecarboxylate (salt) (9CI), Nicotinic acid, compd. with 7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]theophylline (1:1) (8CI), Theophylline, 7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-, mononicotinate (salt) (8CI), Theophylline, 7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-, nicotinate (6CI), 7-[2-Hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]theophylline nicotinate, 7-[2-Hydroxy-3-[N-(2-hydroxyethyl)-N-methylamino]propyl]-1,3-dimethylxanthine pyridine-3-carboxylate, 7-[2-Hydroxy-3-[N-methyl-N-(2-hydroxyethyl)amino]-propyl]-theophylline nicotinate, 7-[3-(Methyl-2-hydroxyethylamino)-2-hydroxypropyl)]theophylline nicotinate, Angioamin, Angiomin, Complamex, Complamin, Complamin retard, Contamex, Landrina 300, NSC 113217, Sadamine, SK 331A, Stenalgil, Teonicol, Vedrin, Xanthinol niacinate, Xanthinol nicotinate, Xantinol nicotinate, Xavin
IUPAC Name 7-[2-hydroxy-3-[2-hydroxyethyl(methyl)amino]propyl]-1,3-dimethylpurine-2,6-dione;pyridine-3-carboxylic acid
SMILES CN(CCO)CC(O)Cn1cnc2N(C)C(=O)N(C)C(=O)c12.OC(=O)c3cccnc3
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C13H21N5O4.C6H5NO2/c1-15(4-5-19)6-9(20)7-18-8-14-11-10(18)12(21)17(3)13(22)16(11)2;8-6(9)5-2-1-3-7-4-5/h8-9,19-20H,4-7H2,1-3H3;1-4H,(H,8,9)
Formula Weight 434.1914
Chemical Name or Material Xanthinol Nicotinate