missing translation for 'onlineSavingsMsg'
Learn More

Sarstedt X500 S-Monovette 2,7ml, 66 x 11mm, Kalium- EDTA .

Brand:  Sarstedt 05.1167

Encompass_Preferred

X500 S-Monovette 2,7ml, 66 x 11mm, Kalium- EDTA .

Product Code. 10207832

  • 98.20 € / Pack of 500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • S-Monovette 2,7ml, 66 x 11mm, Kalium- EDTA .

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Ethylenediaminetetraacetic acid tripotassium salt dihydrate

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.