Learn More
X5 5ML STANDARD BULB 65 %HR
Brand:  Rotronic EA65SCS
X5 5ML STANDARD BULB 65 %HR
Additional Details : Weight : 0.80000kg
Product Code. 11920665
  • 142.00 € / Pack of 5
Estimated Shipment: 21-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • humidity standard 65%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P301-IF SWALLOWED:.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contains : lithium chloride