Learn More
Hilgenberg X100 Glass capillaries borosilicate glass 3.3, filament, cut ends L=100±0.5 Da=1±0.05 Di=0.58
Brand:  Hilgenberg 1403547
X100 Glass capillaries borosilicate glass 3.3, filament, cut ends L=100±0.5 Da=1±0.05 Di=0.58
Product Code. 18832012
  • 417.02 € / Pack of 100
Only null left
Estimated Shipment: 05-02-2026