missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Plate Count Skim Milk Agar
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
Description
Achieve accurate microbial enumeration in dairy products with Plate Count Skim Milk Agar, providing reliable results for quality control.
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2°C to 12°C |
| Format | 90 mm Plate |
| Manufacturing Origin | Germany |
| Product Type | Agar Plate |
| Fill Weight | 17 g |
| Quantity | 10 Plates |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction