Learn More
Klever X-Change Plus (Wider Cut Head with Scraper)
Recessed blade of X-Change Plus (Wider Cut Head with Scraper) reduces cut injuries and damaged goods.
829.98 €
Specifications
Blade Material | Plastic Polymer |
---|---|
For Use With (Application) | Cuts Boxes, Film, Tape, and Thousands More Applications |
Disposable | No |
Sterility | Non-sterile |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
17709027
|
Klever
PLS-200XC-35G |
Orange |
829.98 €
Pack of 50 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
17719027
|
Klever
PLS-200XC-35W |
White |
829.98 €
Pack of 50 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
17729027
|
Klever
PLS-200XC-35Y |
Yellow |
829.98 €
Pack of 50 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
Description
- Cuts boxes, film, tape and thousands more applications.
- Lighter weight, multiple-position handle for control and reach Innovative tape splitter.
- Adavnced plastic polymers.
- Highest safety level.
- Interchangeable, replaceable head.
Specifications
Plastic Polymer | |
Cuts Boxes, Film, Tape, and Thousands More Applications | |
No | |
Non-sterile |