Learn More
WZ-3146, MedChemExpress
Product Code. 16447657
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16447657
Brand: Medchem Express HY12001
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress WZ3146 is a mutant selective EGFR inhibitor with IC50s of 2, 2, 5, 14 and 66 nM for EGFRL858R, EGFRL858R/T790M, EGFRE746_A750, EGFRE746_A750/T790M and EGFR, respectively.
WZ3146 is a mutant selective EGFR inhibitor with IC50s of
Specifications
Chemical Name or Material WZ-3146
CAS 1214265-56-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H25ClN6O2
Quantity 5 mg
Solubility Information DMSO : 62.5 mg/mL (134.42 mM; ultrasonic and warming and heat to 60°C)
SMILES CN1CCN(C2=CC=C(C=C2)NC3=NC=C(C(OC4=CC(NC(C=C)=O)=CC=C4)=N3)Cl)CC1
Molecular Weight (g/mol) 464.95
Formula Weight 464.95
Percent Purity 99.11%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease