missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Verenigde Kunststof Bedrijven™ Wrench for Cap DIN 51
Description
- WRENCH DIN 51
- Color Black
- Effortless safe and secure
- Indispensable tool
Specifications
Specifications
| Material | HDPE |
| Color | Black |
| For Use With (Application) | Jerry Can |
| No. per Pack | 1 Ea. |
| Product Type | Wrench |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?