missing translation for 'onlineSavingsMsg'
Learn More

Reitenspiess™ Wooden-handled Drinking Glass Brush

Product Code. p-7162428
Click to view available options
Width (Metric) Bristle:
80 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10007701

Brand: Reitenspiess™ 11780002

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Black horsehair-bristled, wooden-handled brush for cleaning drinking glasses.

  • Perfectly shaped for cleaning drinking glasses.
  • Constructed of wood and black horsehair.

Specifications

Product Type Drinking Glass Brush
Bristle Material Horse Hair
Handle Material Wood
Length (Metric) 390 mm
Length (Metric) Brush 120 mm
Width (Metric) Bristle 80 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.