missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WNT7B Polyclonal Antibody
GREENER_CHOICE

Product Code. 16619934
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16619934 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16619934 Supplier Invitrogen™ Supplier No. PA5103480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total WNT7B.

Wnt7b encodes a member of the WNT gene family, which consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. Among members of the human WNT family, this gene product is most similar to WNT7A protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT7B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Wnt7b
Gene Accession No. P28047, P56706
Gene Alias Protein Wnt-7b; wingless-related MMTV integration site 7B; wingless-type MMTV integration site family member 7B; wingless-type MMTV integration site family, member 7B; Wnt family member 7B; WNT7B; Wnt-7b
Gene Symbols Wnt7b
Host Species Rabbit
Immunogen A synthesized peptide derived from human WNT7B(Accession P56706), corresponding to amino acid residues N276-C326.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22422, 315196, 7477
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.