Learn More
PeproTech WNT3A Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Brand: PeproTech 500-P251-1MG
Description
Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Wnt-3a. Anti-Human Wnt-3a-specific antibody was purified by affinity chromatography employing an immobilized Human Wnt-3a matrix. Sandwich ELISA: To detect Human Wnt-3a by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human Wnt-3a (500-P251Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Wnt-3a. Western Blot: To detect Human Wnt-3a by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Wnt-3a is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Wnt3a, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
Specifications
| WNT3A | |
| Polyclonal | |
| Unconjugated | |
| WNT3A | |
| MGC119418; MGC119419; MGC119420; Protein Wnt-3a; vestigial tail; vt; wingless-related MMTV integration site 3A; wingless-type MMTV integration site family member 3A; wingless-type MMTV integration site family, member 3 like; wingless-type MMTV integration site family, member 3A; Wnt 3a; Wnt family member 3A; wnt[a]; wnt3; wnt3 l; wnt3 like; Wnt3a; Wnt-3a; WNT3A protein; wnt3l; Zwnt[a] | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 89780 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P56704 | |
| WNT3A | |
| E.coli-derived Recombinant Human Wnt-3a. | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.