missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WNT11 Polyclonal Antibody

Product Code. 15365201
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15365201 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15365201 Supplier Invitrogen™ Supplier No. PA534317

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WNT11 Polyclonal Antibody for IHC (P)

Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene WNT11
Gene Accession No. O96014, P48615, P49339
Gene Alias HWNT11; Protein Wnt-11; wingless-related MMTV integration site 11; wingless-type MMTV integration site family, member 11; Wnt family member 11; Wnt11; Wnt-11
Gene Symbols WNT11
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from internal region of human WNT11.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009030, 100064184, 100521912, 100774690, 140584, 22411, 395562, 485183, 613288, 7481
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Chicken, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.