Learn More
Wuertt Celluloid™ Wire Beschic Test Tube Rack
Specifications
Specifications
| Material | Wire |
| Positions | 12 |
| For Use With (Application) | 4 x 6 Glasses, nylon |
| Quantity | 1 Each |
| Temperature (Metric) Range | 80°C |
| Dimensions (L x W x H) | 50 x 130 x 70 mm |
| Length (Metric) | 50 mm |
| Width (Metric) Slide | 130 mm |
| Product Type | Test Tube Rack |
| Height (Metric) | 70 mm |
| Show More |