Learn More
Tork Wiping Paper Plus W1
Product Code. p-200113529 Shop All Tork Products
Change view
Click to view available options
Color:
Blue
White
Unit Size:
1 roll
6 rolls
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30057816 Blue 1 roll
30243786 White 6 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30057816 Supplier Tork Supplier No. 130050
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The 2 ply multi-purpose Tork Wiping Paper Plus is ideal for mopping up liquids and for hand drying.
  • Wipe dirty hands and lightly soiled surfaces with ease with our Tork Wiping Paper Plus.
  • Mop up spills easily, due to the high absorbency of this wiping paper.
  • Tackle tasks requiring strength and absorbency in no time with this multipurpose design.
  • Swiftly carry and dispose of packaging with Tork Easy Handling™ Plastic Packaging.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wiping Paper
Length (English) 557.74 yd.
Length (Metric) 510 m
Width (English) 14.53 in.
Width (Metric) 36.9 cm
Material Paper
Certifications/Compliance EU Ecolabel, ISO 50001, FSC Chain-Of-Custody, OHSAS 18001, HACCP, EMAS, FSC Mix Virgin and Rec.fibre, ISO 9001, ISO 14001
Color Blue
Ply 2-ply
Certifications/Compliance (Europe) EU Ecolabel, ISO 50001, FSC Chain-Of-Custody, OHSAS 18001, HACCP, EMAS, FSC Mix Virgin and Rec.fibre, ISO 9001, ISO 14001