Learn More
TERUMO Winged Infusion Safety Sets SURFLO™
Product Code. 17162824
Change view
Click to view available options
Needle Gauge:
18 G
19 G
21 G
22 G
23 G
25 G
Length (Metric) Tube:
9 cm
30 cm
Unit Size:
Pack of 50
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Length (Metric) Tube unitSize
17162824 22 G 30 cm Pack of 50
17112824 18 G 30 cm Pack of 50
17122824 19 G 30 cm Pack of 50
17132824 21 G 9 cm Pack of 50
17142824 21 G 30 cm Pack of 50
17152824 22 G 9 cm Pack of 50
17172824 23 G 9 cm Pack of 50
17182824 23 G 30 cm Pack of 50
17192824 25 G 30 cm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 17162824 Supplier TERUMO Supplier No. 8SVS22NL30
Please to purchase this item. Need a web account? Register with us today!
Edge
This item is not returnable. View return policy
Intended to introduce or withdraw fluids from the vascular system after vein puncture (18 - 25 G).
  • Made out of kink resistant material to prevent kinking during placement for a higher patient comfort
  • The wings are large and flexible for a good grip and secure fixation
  • Reduced pressure on the blood vessel wall due to slightly angled wings
  • Easy activation of the safety mechanism, fingers tips stay away from the cannula at all times
  • Audible sound during activation of the safety mechanism
TRUSTED_SUSTAINABILITY
Specifications
Product Type Infusion Set
Color Black
Material PVC Tubing, Siliconized Needle
Needle Gauge 22 G
Sterility Sterile
Latex Free Yes
Length (Metric) Tube 30 cm