Learn More
Wighteone, TRC
Specifications
Specifications
| Chemical Name or Material | Wighteone |
| CAS | 51225-30-0 |
| Molecular Formula | C20H18O5 |
| Quantity | 10 mg |
| Synonym | 5,7-Dihydroxy-3-(4-hydroxyphenyl)-6-(3-methyl-2-buten-1-yl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-6-(3-methyl-2-butenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-6-(3-methyl-2-buten-1-yl)-4H-1-benzopyran-4-one, 5,7,4'-Trihydroxy-6-prenylisoflavone, Erythrinin B |
| InChI Formula | InChI=1S/C20H18O5/c1-11(2)3-8-14-16(22)9-17-18(19(14)23)20(24)15(10-25-17)12-4-6-13(21)7-5-12/h3-7,9-10,21-23H,8H2,1-2H3 |
| SMILES | CC(=CCc1c(O)cc2OC=C(C(=O)c2c1O)c3ccc(O)cc3)C |
| IUPAC Name | 5,7-dihydroxy-3-(4-hydroxyphenyl)-6-(3-methylbut-2-enyl)chromen-4-one |
| Molecular Weight (g/mol) | 338.35 |
| Formula Weight | 338.1154 |