Learn More
Motic™ Widefield Eyepiece N-WF15X/F.N.16 with Diopter Adjustment (ESD)
Description
- Widefield style and diopter adjustment provides improved field of view and greater eye relief.
- Magnification: 10X.
- Field number: 16.
Specifications
Specifications
| Type | Widefield Eyepiece |
| Description | F.N.: 16 |
| For Use With (Equipment) | SM-171, GM Series Microscopes |
| Includes | Diopter adjustment, ESD electrostatic shock protection |
| Magnification Power | 15X |