Learn More
IKA™ WiCo RET Control-visc
Shop All IKA Products
Click to view available options
Dimensions (L x W x H):
105 x 160 x 40 mm
Unit Size:
Each
Description
- The WiCo is equipped with a USB socket (Universal Serial Bus) with which the WiCo can be connected with station, the USB icon appears on the screen
- If the WiCo is not connected with station via a USB cable, the data exchange between the station and the WiCo via BluetoothR
- In this case, the BluetoothR icon is displayed
- Depending on the structure of the building, the WiCo can be operated at a distance up to 15 m from the station, using the BluetoothR connection
Specifications
Specifications
| Temperature (Metric) | 5 to 40°C |
| Type | Magnetic Stirrer Remote |
| Weight (Metric) | 0.3 kg |
| Dimensions (L x W x H) | 105 x 160 x 40 mm |