missing translation for 'onlineSavingsMsg'
Learn More

WIBG Monoclonal Antibody (OTI11C5), TrueMAB™, OriGene

Product Code. 17596543
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17596543 30 μL 30µL
17576543 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17596543 Supplier Origene Technologies Supplier No. TA806390S
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

WIBG Monoclonal antibody specifically detects WIBG in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen WIBG
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI11C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Tnfrsf18
Gene Accession No. Q9Y5U5
Gene Alias A030010B05Rik; Partner of Y14 and mago; protein wibg homolog; PYM; PYM homolog 1 exon junction complex-associated factor; PYM homolog 1, exon junction complex associated factor; Pym1; RGD1306096; WIBG; within bgcn homolog; within bgcn homolog (Drosophila)
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 26-162 of human TNFRSF18 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8784
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.