Learn More
DWK Life Sciences Wheaton™ Shorty Vials™ Glass Sample Vials in Lab File™: with Caps
Product Code. 12424991 Shop All Wheaton Science Products Products
Click to view available options
Volume (Metric):
2 mL
4 mL
6 mL
Diameter (Metric) Outer:
15 mm
17 mm
19 mm
Material (Liner):
14B Rubber
PTFE/14B Rubber
Unit Size:
Pack of 200
Product Code. 12424991
Brand: DWK Life Sciences W225292
Only null left
Add to basket
Add to basket
Come in convenient Lab File partitioned trays
Offering a variety of uses ranging from sample storage to freeze-drying.
  • Clear vials made from borosilicate glass
  • Fabricated from glass tubing for uniform sidewall and bottom thickness
  • Shorty Vials feature a low profile for greater stability and less dead volume
  • Supplied with attached caps to help maintain cleanliness or without caps (replacement caps can be purchased separately)
  • Vials and caps are autoclavable
  • Selected items fit M-T Vial File™—polystyrene trays with partitions that provide an easy way to inventory samples or to store empty vials
Clear Vials
  • Made from low extractable borosilicate glass to provide superior chemical resistance
  • Conform to USP Type I and ASTM E 438 Type I, Class A requirements
Cap Liners
  • 14B rubber-lined black phenolic screw cap for aqueous samples
  • PTFE/14B rubber-lined black phenolic screw cap for organic samples
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) 14B Rubber
Color Clear
Diameter (Metric) Outer 17 mm
Material Borosilicate Glass (Low Extractable)
Volume (Metric) 4 mL
For Use With (Application) For uses ranging from sample storage to freeze drying
Thread Style Screw
Volume (English) 1.08 dr.
Closure Material Phenolic
Autoclavable Autoclavable
Closure Type Screw Cap
Height (Metric) 41 mm
Certifications/Compliance ASTM E438, USP Type I, Type I, Class A
For Use With (Equipment) Vial Rack