Learn More
DWK Life Sciences™ Wheaton™ Polyacetal Clamps
Product Code. p-31190 Shop All Wheaton Science Products Products
Click to view available options
Color:
Blue
Dark Green
Yellow
Unit Size:
Pack of 12
Product Code. 12438922
Brand: DWK Life Sciences™ 297746
Please to purchase this item. Need a web account? Register with us today!
Color-coded clamps hold standard taper joints securely under moderate pressure
  • Molded from polyoxymethylene (POM), an acetal resin
  • Will not scratch glass and can withstand temperatures up to 150°C
  • Resistant to bases and weak acids
  • Not resistant to halogen gases and concentrated acids
TRUSTED_SUSTAINABILITY
Specifications
Type Standard Taper Clamp
Material POM Acetal
For Use With (Equipment) Kuderna Danish Evaporative Concentrator
Color Yellow