Learn More
DWK Life Sciences Wheaton™ I-LOC Closures
Shop All Wheaton Science Products Products
Click to view available options
Color:
Black
Blue
Red
White, Gray
Yellow
Unit Size:
Pack of 100
Description
Eliminate use of separate aluminum seal and rubber stopper
- Polypropylene closures
- Gray bromobutyl stoppers
Specifications
Specifications
| Material (Liner) | Bromobutyl |
| Color | Yellow |
| Material | Polypropylene |
| Autoclavable | Autoclavable |
| Type | Screw Cap |
| Closure Size | 20-400 |