Learn More
DWK Life Sciences Wheaton™ Black Phenolic Screw Caps with PTFE faced 14B Rubber Liners for E-Z Ex-Traction™ Vials
Product Code. 10140203 Shop All Wheaton Science Products Products
Click to view available options
For Use With (Equipment):
10 mL Vial
2 and 4mL Vial
20mL Vial
8mL Vial
Neck Finish:
13-425
15-425
22-400
24-400
No. per Case:
100 Cs.
1000 Cs.
250 Cs.
500 Cs.
Unit Size:
Pack of 100
Pack of 1000
Pack of 250
Pack of 500
Product Code. 10140203
Brand: DWK Life Sciences W240823
Only null left
Add to basket
Add to basket
Designed to ensure the ultimate in product safety
  • Autoclavable
  • Liner made of styrene-butadiene rubber faced with PTFE
  • Excellent chemical resistance
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Phenolic Resin
No. per Case 1000 Cs.
Neck Finish 15-425
Certifications/Compliance ISO 9001
For Use With (Equipment) 8mL Vial
Closure Liner PTFE/Rubber