Learn More
DWK Life Sciences Wheaton™ Black Phenolic Solid Top Screw Caps
Product Code. 10423332 Shop All Wheaton Science Products Products
Click to view available options
Closure Size:
15-425
18-400
20-400
24-400
Unit Size:
Pack of 144
Pack of 200
Pack of 500
Product Code. 10423332
Brand: DWK Life Sciences 240215
Only null left
Add to basket
Add to basket
Providing safe, secure and chemically inert storage. Wheaton™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.
Providing safe, secure and chemically inert storage. Wheaton™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.
  • 14B rubber lined caps feature a nontoxic white styrene-butadiene rubber liner ideal for cell culture work
  • Aluminum foil faced pulp board liners can be used with organic solvents
  • Use LDPE lined caps with distilled water, analytical standards and reagents
TRUSTED_SUSTAINABILITY
Specifications
Color Black
For Use With (Application) Ideal for cell culture work
Material Phenolic
Liner Material Styrene Butadiene Rubber
Product Type Screw Cap
Closure Size 18-400
Autoclavable Autoclavable