missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polycap Disposable Capsules, 75 AS

Product Code. 11324670
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Inlet Style:
Female National Pipe Thread
Stepped Barb
Stepped Barb with filling bell
Outlet Style:
Female National Pipe Thread
Stepped Barb
Stepped Barb with filling bell
Sterility:
Non-sterile
Sterile
Unit Size:
Each
Pack of 5

Product Code. 11324670

Brand: Cytiva 67067502

Please to purchase this item. Need a web account? Register with us today!

Polycap 75 AS filters for filtration of aqueous solutions

  • Sterile and nonpyrogenic
  • Integrity testable
  • With inlet vent
  • Filter Media: Low-extractable, hydrophilic nylon membrane and glass microfiber prefilter

  • Filling bell model (09-744-61) minimizes contamination when filtering into bottle or flask

Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Material (Membrane) Nylon
Pore Size 0.2 μm
Sterility Sterile
Filtration Area 820 cm2
For Use With (Application) Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
Housing Material Polypropylene
Inlet Style Stepped Barb with filling bell
Outlet Style Stepped Barb with filling bell
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.