Learn More
Cytiva Whatman™ Grade 470
Click to view available options
Dimensions (L x W):
203 x 305 mm
450 ft x 1.5 in.
460 x 570 mm
Unit Size:
Each
Pack of 100
Pack of 25
Description
- Soft surface
- For gelatinous samples
- For the absorption of culture media
- Used as blotting paper and amino acid chromatography
Specifications
Specifications
| Format | Roll |
| For Use With (Application) | Electrophoresis and amino acid chromatography |
| Grade | 470 |
| Dimensions (L x W) | 450 ft x 1.5 in. |
| Length (Metric) | 137.1 m |
| Width (Metric) | 38 mm |