missing translation for 'onlineSavingsMsg'
Learn More

DiversifiedBiotech Well Orienters

Product Code. 30282291
Change view
Click to view available options
Product Type:
Well Orienter
Well Orienter Stickers
Unit Size:
Pack of 4
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
30282291 Well Orienter Stickers Pack of 50
30282283 Well Orienter Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30282291 Supplier DiversifiedBiotech Supplier No. WEST1000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For easy pipetting and identification with mutliple microplates

  • For Easy Pipetting and Identification of Samples in Microplates
  • A thin, plastic card that slips under the well plate, providing sharp, alpha-numeric characters to identify each sample in a 96 - 48 or 24 - well microplate
  • Permanently label mutliple microplates with clear Well Orienter Stickers
  • Each well is clearly marked with its corresponding row number and column letter for easy cross-referencing
  • The paper backing is scored in the middle allowing for easy aligning on the underside of the plate

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Color Black, Clear Bottom
Disposable Single-use
Product Type Well Orienter Stickers
For Use With (Equipment) Well Plates
Material Plastic
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round
For Use With (Application) Cross Referencing
Includes Well Orienter Only
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.