missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Welder RF Gloves

Product Code. 12691113
Click to view available options
:
Size: 8
This item is not returnable. View return policy

Product Code. 12691113

Brand: Honeywell Safety Products™ 205403008

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Especially suitable for heavy handling and ARC and MIG welding operations, these gloves are constructed of heat-resistant split leather. Honeywell Safety Products™ Welder RF Gloves are also stitched with Kevlar™ thread for additional durability and heat resistance.

  • Suitable for ARC and MIG welding.
  • Heat-resistant split leather back.
  • Grain leather palm for mechanical resistance.
  • Grain leather reinforcement between the thumb and index finger
  • Gunn cut, winged thumb, reinforced index finger, Crispin type.
  • Split leather cuff to protect the forearm.
  • Stitched with sturdy and heat-resistant Kevlar™ thread.
  • U.S. assembly.
TRUSTED_SUSTAINABILITY

Specifications

Back Material Split Leather
Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 407-2004 Heat hazards, EN 12 477-2001 Welding hazards type A, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number 0075/797/162/01/06/0353, EC Attestation Number 0075/797/162/01/06/0353
Classification Dexterity: 5
Cuff Material Split Leather
Cut Style Gunn
Cut Resistance Rating 1
Material Split Leather, Leather
Product Type Welding Gloves (Type A)
Size 8
Style Gauntlet
Thickness (Metric) 1.1 to 1.3 mm
For Use With (Application) Automotive, Building, Construction, Iron, Steel, Ship Building and Welding industries.
Includes Reinforced thumb and forefinger in grain leather.
Palm Type Grain Leather
Puncture Rating 2
Quantity 10Pair/PK
Resistant To Heat
Tear Rating 3
Thumb Style Wing
Length (Metric) 40 cm
Length (Metric) Cuff 20 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.