Learn More
Tocris Bioscience™ WDR5 0103
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
WDR5 0103 is a WD repeat-containing protein 5 (WDR5) antagonist (Kd = 450 nM). Disrupts WDR5 interaction with MLL and inhibits MLL core complex methyltransferase activity in vitro. Has no effect on a panel of seven other methyltransferases, including SETD7.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | WDR5 0103 |
| Purity | >99% |
| Molecular Weight (g/mol) | 383.44 |
| Inhibitors | WDR5 0103 |