missing translation for 'onlineSavingsMsg'
Learn More

WAY-600, MedChemExpress

Product Code. 16450236
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16450236

Brand: Medchem Express HY15272

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WAY-600 is a potent, ATP-competitive, and selective mTOR inhibitor with an IC50 of 9 nM for recombinant mTOR enzyme. WAY-600 blocks mTOR complex 1/2 (mTORC1/2) assemble and activation.

WAY-600 is a potent, ATP-competitive, and selective mTOR inhibitor with an IC50 of 9 nM for recombinant mTOR enzyme. WAY-600 blocks mTOR complex 1/2 (mTORC1/2) assemble and activation.

Specifications

Chemical Name or Material WAY-600
CAS 1062159-35-6
Color White
Purity Grade Notes Research
Molecular Formula C28H30N8O
Quantity 2 mg
Solubility Information DMSO : 50 mg/mL (101.09 mM; Need ultrasonic)
SMILES C1(C=CN2)=C2C=CC(C3=NC(N(C4CCN(CC5=CC=CN=C5)CC4)N=C6)=C6C(N7CCOCC7)=N3)=C1
Molecular Weight (g/mol) 494.59
Formula Weight 494.59
Percent Purity 95.12%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.