Learn More
BRAND™ Water Still Hosecock Clamp
Product Code. 11613249 Shop All Brand Products
Click to view available options
:
Tubing Size: Up to 14 mm outer diameter
Product Code. 11613249
Brand: BRAND™ 584700
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Nickel plated brass for tubes up to 14 mm outer diameter
TRUSTED_SUSTAINABILITY
Specifications
Material Nickle Plated Brass
For Use With Water Still
Tubing Size Up to 14 mm outer diameter