Learn More
Miele Professional™ Water purification
Product Code. 15410535 Shop All Miele Products
Change view
Click to view available options
For Use With (Equipment):
Washer-disinfectors
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15410535 Washer-disinfectors Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15410535 Supplier Miele Professional™ Supplier No. 9782600
Please to purchase this item. Need a web account? Register with us today!
Quick connection (SK) for the simple and convenient changing of demineralisation cartridges
  • 2 x VA plug connection 3/4'', 2 x quick connection 3/4''
  • Including Seals for the hose attachment
TRUSTED_SUSTAINABILITY
Specifications
Color (Overall) Stainless Steel
For Use With (Equipment) Washer-disinfectors
Type Water Pre-treatment
Material Stainless Steel