Learn More
WTW™ Water Hardness Test Kits
Click to view available options
Type:
RH Residual Hardness
Water Hardness, Total
Unit Size:
25g
Pack of 25
Specifications
Specifications
| Analytes | Phthalein complexone |
| For Use With (Application) | To check of ion-exchanger plants for the production of deionized water |
| For Use With (Equipment) | PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers |
| Includes | Certificates |
| No. of Tests | 25 |
| Range | 0.075 to 0.750 °d, 0.5 to 5 mg/L Ca |
| Sufficient For | 25 Tests |
| Test Method | Photometric |
| Type | RH Residual Hardness |
Safety and Handling
Product Identifier
- residual hardness cell test method
- Warning
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : sodium hydroxide