Learn More
Heidolph™ Water Distribution Manifold
Shop All Heidolph Instruments Products
Click to view available options
For Use With (Equipment):
Starfish Magnetic Stirrer System.
Unit Size:
Each
Description
- Distribute to up to 5 condensers.
- Recombines flow to one outlet pipe.
- Leak-proof shut-off valves.
Specifications
Specifications
| For Use With (Equipment) | Starfish Magnetic Stirrer System. |
| For Use With (Application) | Water Distribution. |
| Includes | Leak-proof shut-off valves. |